The Micro BCA Reagent C (MC) is a unique copper sulfate solution. This product is sufficient for 3,200 microplate assaysRead more
Have Questions?
Catalog Number
Quantity
23234
12 mL
Catalog number 23234
Price (USD)
53.75
Each
Add to cart
Quantity:
12 mL
Request bulk or custom format
Price (USD)
53.75
Each
Add to cart
The Micro BCA Reagent C (MC) is a unique copper sulfate solution. This product is sufficient for 3,200 microplate assays when mixed with Reagents MA and MB.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayBCA Assay
DescriptionPierce Micro BCA Reagent C (MC)
For Use With (Application)Solution-based Detection, Absorbance
For Use With (Equipment)Spectrophotometer, Microplate Reader
Product LineMicro BCA™, Pierce™
Product TypeProtein Quantitation Assay
Quantity12 mL
SpecificityNot Target-Specific
Sufficient For3200 Microplate Assays
Detection MethodColorimetric
Unit SizeEach
Contents & Storage
Upon receipt store product at room temperature. Product shipped at ambient temperature.
Frequently asked questions (FAQs)
Can you provide the shelf-life for the Pierce Micro BCA Protein Assay Kit?
The Pierce Micro BCA Protein Assay Kit is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.