Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Create a compact, space saving, reagent dispensing bank using chemical-resistant Thermo Scientific™ Nalgene™ Low-Profile autoclavable polypropylene copolymer (PPCO) Lowboys. The low-profile design of these high density polyethylene lowboys fits into tight spaces on shelves and in refrigerators and can stack three high. Chemically compatible with a wide range of lab liquids. Convenient integral carry handle for safe, easy transport. Nalgene polypropylene copolymer carboys can be autoclaved before refilling to eliminate bacterial growth when storing sterile solutions. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables.
Closure Size | 63 mm |
Type | Carboy with Spigot |
Description | Autoclavable Lowboy w/Spigot PP 8 L |
Material | PPCO |
Capacity (English) | 2 gal. |
Capacity (Metric) | 8 L |
Length (Metric) | 305 mm |
Diameter (English) Inner Neck | 1.73 in. |
Diameter (Metric) Inner Neck | 44 mm |
Width (Metric) | 366 mm |
Height (English) With Closure | 6.26 in. |
Height (Metric) With Closure | 159 mm |
No. per Case | 4 |
No. per Pack | 1 |
Unit Size | Each |
Closure Size | 63 mm |
Type | Carboy with Spigot |
Description | Autoclavable Lowboy w/Spigot PP 8 L |
Material | PPCO |
Capacity (English) | 2 gal. |
Capacity (Metric) | 8 L |
Length (Metric) | 305 mm |
Diameter (English) Inner Neck | 1.73 in. |
Diameter (Metric) Inner Neck | 44 mm |
Width (Metric) | 366 mm |
Height (English) With Closure | 6.26 in. |
Height (Metric) With Closure | 159 mm |
No. per Case | 4 |
No. per Pack | 1 |
Unit Size | Case of 4 |
Closure Size | 63 mm |
Type | Carboy with Spigot |
Description | Autoclavable Lowboy w/Spigot PP 15 L |
Material | PPCO |
Capacity (English) | 4 gal. |
Capacity (Metric) | 15 L |
Length (Metric) | 371 mm |
Diameter (English) Inner Neck | 1.73 in. |
Diameter (Metric) Inner Neck | 44 mm |
Width (Metric) | 371 mm |
Height (English) With Closure | 8.27 in. |
Height (Metric) With Closure | 210 mm |
No. per Case | 4 |
No. per Pack | 1 |
Unit Size | Case of 4 |
Closure Size | 63 mm |
Type | Carboy with Spigot |
Description | Autoclavable Lowboy w/Spigot PP 15 L |
Material | PPCO |
Capacity (English) | 4 gal. |
Capacity (Metric) | 15 L |
Length (Metric) | 371 mm |
Diameter (English) Inner Neck | 1.73 in. |
Diameter (Metric) Inner Neck | 44 mm |
Width (Metric) | 371 mm |
Height (English) With Closure | 8.27 in. |
Height (Metric) With Closure | 210 mm |
No. per Case | 4 |
No. per Pack | 1 |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Closure Size | Capacity (Metric) | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
2324-0008PK also known as 2324-0008 | Each | 63 mm | 8 L | 316.80 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2324-0008 | Case of 4 | 63 mm | 8 L | 855.00 Online price 1,074.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2324-0015 | Case of 4 | 63 mm | 15 L | 937.31 Online price 1,240.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2324-0015PK also known as 2324-0015 | Each | 63 mm | 15 L | 365.90 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Includes:
Polypropylene screw closures.
.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.