Pierce™ Modified Lowry Protein Assay Kit
Pierce™ Modified Lowry Protein Assay Kit
Thermo Scientific™

Pierce™ Modified Lowry Protein Assay Kit

The Pierce Modified Lowry Protein Assay is a stable form of a traditional, two-component, folin phenol- and copper-based reagent systemRead more
Have Questions?
Catalog NumberQuantity
23240530 mL
Catalog number 23240
Price (EUR)
249,00
Each
-
Add to cart
Quantity:
530 mL
Request bulk or custom format
Price (EUR)
249,00
Each
Add to cart
Ask our AI about this Product
The Pierce Modified Lowry Protein Assay is a stable form of a traditional, two-component, folin phenol- and copper-based reagent system to measure total protein concentration compared to a protein standard.

See all available protein assays ›

Features of Modified Lowry Protein Assay include:
Popular method—widely cited in protein research literature
Colorimetric—measure with a standard spectrophotometer or plate reader at 750 nm
Stability—uses a modified cupric sulfate-tartrate reagent that is stable at room temperature
Assay range—exhibits good linearity in the range 1 to 1500 μg/mL (tested with BSA protein)

The Modified Lowry Protein Assay Kit combines a stabilized formulation of the original Lowry reagents and the essential Folin-Ciocalteu Phenol reagent in a complete kit for accurately determining protein concentration in a variety of samples types. Although newer protein assay methods provide greater speed and convenience, the Lowry method remains a popular, accurate, and useful option for many applications.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayLowry Assay
For Use With (Application)Solution-based Detection, Absorbance
For Use With (Equipment)Spectrophotometer, Microplate Reader
Product LinePierce™
Product TypeProtein Quantitation Assay
Quantity530 mL
SpecificityNot Target-Specific
Sufficient For480 Tube Assays or 2400 Microplate Assays
Detection MethodColorimetric
Unit SizeEach
Contents & Storage
Sufficient For: 480 tube assays or 2400 microplate assays

• Modified Lowry Protein Assay Reagent, 480 mL
• 2N Folin-Ciocalteu Phenol Reagent, 50 mL
• Albumin Standard Ampules, 2 mg/mL, 10 x 1 mL

Store at 4°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection