Pierce™ Colorimetric Protease Assay Kit
Pierce™ Colorimetric Protease Assay Kit
Thermo Scientific™

Pierce™ Colorimetric Protease Assay Kit

The Thermo Scientific Pierce Colorimetric Protease Assay Kit measures total protease activity in samples, providing a means to assess theRead more
Have Questions?
Catalog NumberQuantity
23263250 Test Kit
Catalog number 23263
Price (USD)
423.65
Online Exclusive
464.00
Save 40.35 (9%)
Each
Add to cart
Quantity:
250 Test Kit
Request bulk or custom format
Price (USD)
423.65
Online Exclusive
464.00
Save 40.35 (9%)
Each
Add to cart
The Thermo Scientific Pierce Colorimetric Protease Assay Kit measures total protease activity in samples, providing a means to assess the progress of protease isolation procedures or quantify protease contamination in biological samples. This kit uses fully succinylated casein as a substrate for the assay. Hydrolysis of this substrate in the presence of protease results in the release of peptide fragments with free amino-terminal groups. These peptides are reacted with trinitrobenzene sulfonic acid (TNBSA), followed by measurement of the absorbance increase that results from the formation of yellow colored TNB-peptide adducts.

Features of the Colorimetric Protease Assay Kit:

Casein substrate—measure activity of any protease that cleaves casein into peptide fragments
Trypsin standard—quantify protease activity relative to trypsin, a universally accepted reference
Sensitive—1000 times more sensitive than assays that use unmodified forms of casein
Simple and fast—test tube and microplate protocols are completed in less than one hour
Homogenous—addition steps only; no separation, transfer or stop steps required
Customizable—easily adapt time, temperature and pH to optimize sensitivity for proteases of interest

Applications:
• Assess functional integrity and overall progress of protease purification
• Quantify protease contamination in protein samples
• Characterize the activity of unusual proteases compared to trypsin
• Evaluate buffer conditions for their affects on protease function and activity

Additional Specifications:
• Substrate: succinylated casein (not available separately)
• Basis of assay: detection of primary amines resulting from proteolysis
• Detection reagent: trinitrobenzene sulfonic acid (TNBSA)
• Measurement: absorbance at 450nm (plate reader or spectrophotometer)

Related Products
Pierce™ Fluorescent Protease Assay Kit
FITC-Casein for Pierce™ Fluorescent Protease Assay Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayProtease Assay
For Use With (Application)Absorbance
For Use With (Equipment)Microplate Reader
Product LinePierce™
Product TypeProtein Activity Assay
Quantity250 Test Kit
SubstrateCasein
Substrate PropertiesProtein-Based Substrate
Substrate TypeProtease Substrate
Sufficient For250 Microplate Assays
TargetProteases
Detection MethodColorimetric
Immunoassay Kit Format96-well Plate
Unit SizeEach
Contents & Storage
Sufficient For: 250 microplate assays using a standard plate reader (absorbance at 450 nm)
• Succinylated Casein, 5 x 10 mg
• TNBSA Solution, 2 mL
• TPCK Trypsin Standard, 50 mg
• Borate Buffer Pack (makes 500 mL), 1 pack

Store at 4°C.
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link