Pierce™ Phosphoprotein Phosphate Estimation Assay Kit
Pierce™ Phosphoprotein Phosphate Estimation Assay Kit
Thermo Scientific™

Pierce™ Phosphoprotein Phosphate Estimation Assay Kit

The Thermo Scientific Phosphoprotein Phosphate Estimation Kit enables classification and identification of proteins as phosphorylated (serine and threonine), as wellRead more
Have Questions?
Catalog NumberQuantity
23270100 mL
Catalog number 23270
Price (USD)
674.00
Each
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)
674.00
Each
Add to cart
Ask our AI about this Product
The Thermo Scientific Phosphoprotein Phosphate Estimation Kit enables classification and identification of proteins as phosphorylated (serine and threonine), as well as semi-quantitative assessment of the phosphorylation level.

Features of the Phosphoprotein Phosphate Estimation Kit:

Specific—measures phosphoserine (p-Ser) and phosphothreonine (p-Thr) only; does not measure phosphotyrosine (p-Tyr)
Convenient—test tube and 96-well microplate protocols included and require less than 90 minutes to perform
Semi-quantitative—calculate moles of phosphate (P) per mole of protein using the included phosvitin standard
Customizable—format is adaptable to development of specific assays for pure phosphoproteins when previously characterized standards are available

The Phosphoprotein Phosphate Estimation Assay Kit is designed to aid in characterization of the status and extent of phosphorylation of purified protein samples. The assay is based on the alkaline hydrolysis of phosphate from seryl and threonyl residues in phosphoproteins and quantification of the released phosphate with malachite green and ammonium molybdate. The assay is easily performed in 96-well microplates or test tubes and is completed in about one hour.

Applications:
• Identify proteins as containing phosphoserine or phosphothreonine phosphorylations
• Estimate the amount of pS and pT phosphorylation
• Develop specific quantitative assays for well-characterized phosphoproteins of interest

The assay can be used to identify whether a purified protein contains either phospho-serine (p-Ser) or phospho-threonine (p-Thr) as well as to estimate the level of this type of phosphorylation. For quantitation, the test protein sample is compared with specific concentrations of phosvitin, a phosphoprotein of known phosphorylation level. The alkaline hydrolysis step does not release phosphate from phospho-tyrosine (p-Tyr) residues in peptide linkage. Therefore, a negative result for an unknown purified protein preparation indicates that the protein is either (1) not a phosphoprotein or (2) is phosphorylated exclusively at tyrosine residues. In the latter case, Western blot analysis using an anti-phosphotyrosine antibody will be necessary to distinguish between these two possibilities.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayPhosphoprotein Phosphate Estimation Assay
For Use With (Application)Solution-based Detection, Absorbance
For Use With (Equipment)Spectrophotometer, Microplate Reader
Product LinePierce™
Product TypePhosphoprotein Phosphate Estimation
Quantity100 mL
SpecificityNot Target-Specific
Sufficient For500 Tube Assays or 1920 Microplate Assays
Detection MethodColorimetric
Unit SizeEach
Contents & Storage
Sufficient For: 500 tube assays or 1920 microplate assays
• Ammonium molybdate Solution, 25 mL
• Malachite Green Solution, 75 mL
• Phosvitin Positive Control, 1 mg
• TBS Pack (makes 500 mL), 1 pack

Upon receipt, store components at room temperature. Once dissolved, store Phosvitin solution at 4°C (or -20°C for long-term storage).
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link