D-Panthenol, 98+%
Thermo Scientific Chemicals

D-Panthenol, 98+%

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 232801000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name(2R)-2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
Molecular FormulaC9H19NO4
InChI KeySNPLKNRPJHDVJA-ZETCQYMHSA-N
SMILESCC(C)(CO)[C@@H](O)C(=O)NCCCO
Molecular Weight (g/mol)205.25
View more
SpecificationsSpecification SheetSpecification Sheet
Assay>=98.0 %
Specific optical rotation+29.0° to +32.0° (20°C, 589 nm) (c=5, H2O)
Appearance (Form)Viscous liquid or solid
Refractive index1.4950 to 1.5020 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only