Thermo Scientific™

Pierce™ Peroxide Assay Reagent A

Catalog number: 23281
Have Questions?
Thermo Scientific™

Pierce™ Peroxide Assay Reagent A

Catalog number: 23281
Have Questions?
Catalog Number
23281
Unit Size
1 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
232811 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce Peroxide Assay Reagent A is a formulation consisting of 25mM ammonium ferrous sulfate. This formulation is one of the component of Quantitative Peroxide Assay Kit used for detecting and measuring hydrogen peroxide levels based on the oxidation of ferrous to ferric ion in the presence of xylenol orange, present in the biological and other liquid samples.

Applications of Pierce Quantitative Peroxide Assay Kits:
• General hydrogen peroxide (H2O2) measurement in protein samples
• Quantitation of peroxides in detergents
• Assessment of the effects of peroxides on cellular activity
• Determination of protein glycation; Measurement of peroxide accumulation in lipids

In these assay kits, hydroperoxides convert the Fe2+ to Fe3+ at acidic pH. With the aqueous-compatible formulation, peroxide first reacts with sorbitol, converting it to a peroxyl radical, which in turn initiates Fe2+ oxidation to Fe3+. In the lipid compatible formulation, the peroxide converts the Fe2+ to Fe3+ directly. In a sulfuric acid solution, the Fe3+ complexes with the xylenol orange (XO) dye to yield a purple product having an absorbance maximum at 560nm. Peroxide levels in test samples can be determined by calculation from the know extinction coefficient of the XO-Fe complex or by reference to a standard curve prepared with hydrogen peroxide solution.Most proteins do not interfere with these assays, although some metal chelators may require use of a blank (i.e., control) to mitigate these effects.




Related Products
Pierce™ Quantitative Peroxide Assay Kit (Aqueous)
Pierce™ Quantitative Peroxide Assay Kit (Lipid)
Pierce™ Peroxide Assay Reagent B
Pierce™ Peroxide Assay Reagent C
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay
Peroxide Assay Reagent A
Detection Method
Colorimetric
Product Line
Pierce™
Product Type
Quantitative Peroxide Assay
Specificity
Not Target-Specific
For Use With (Application)
Solution-based Detection, Absorbance
For Use With (Equipment)
Spectrophotometer, Microplate Reader

Contents & Storage

Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text