Thermo Scientific Chemicals

Hexamethylenetetramine, 98.5%, contains an anticaking agent, Thermo Scientific Chemicals

Catalog number: 232890025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Hexamethylenetetramine, 98.5%, contains an anticaking agent, Thermo Scientific Chemicals

Catalog number: 232890025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 232890025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
100-97-0
IUPAC Name
1,3,5,7-tetraazatricyclo[3.3.1.1³,⁷]decane
Molecular Formula
C6H12N4
InChI Key
VKYKSIONXSXAKP-UHFFFAOYSA-N
SMILES
C1N2CN3CN1CN(C2)C3
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Water
=<0.5 % (K.F.)
Ash
0.5 % to 3.0 % (anticaking agent)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text