Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | Quantitative Fluorometric Peptide Assay |
For Use With (Equipment) | Microplate Reader |
Product Type | Peptide Assay |
Chemical Reactivity | Amine |
Product Line | Pierce™ |
Starting Material | Peptides, Protease-digested Protein |
Final Product Type | Peptides |
Detection Method | Fluorescent |
Format | Kit |
Workflow Step | In-Process Testing |
Quantity | 500 Assays |
Unit Size | 500 assays |
Description | Quantitative Colorimetric Peptide Assay |
For Use With (Equipment) | Microplate Reader |
Product Type | Peptide Assay |
Product Line | Pierce™ |
Starting Material | Peptides, Protease-digested Protein |
Final Product Type | Peptides |
Detection Method | Colorimetric |
Format | Kit |
Workflow Step | In-Process Testing |
Quantity | 500 Assays |
Unit Size | 500 assays |
Description | Peptide Digest Assay Standard (1 mg/mL) |
For Use With (Equipment) | Microplate Reader |
Product Type | Peptide Digest Assay Standard |
Product Line | Pierce™ |
Starting Material | Peptides, Protease-digested Protein |
Detection Method | Colorimetric, Fluorescent |
Workflow Step | In-Process Testing |
Quantity | 1.5 mL |
Form | Liquid |
Unit Size | 1.5 mL |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
23290 | 500 assays | Quantitative Fluorometric Peptide Assay | 450.65 Online price 504.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
23275 | 500 assays | Quantitative Colorimetric Peptide Assay | 411.65 Online price 463.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
23295 | 1.5 mL | Peptide Digest Assay Standard (1 mg/mL) | 174.65 Online price 183.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|