Ethyl propionylacetate, 98%, Thermo Scientific Chemicals
Ethyl propionylacetate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl propionylacetate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
Packaging:
Glass bottle
Catalog number 232910250
Price (USD)/ Each
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4949-44-4
IUPAC Nameethyl 3-oxopentanoate
Molecular FormulaC7H12O3
InChI KeyUDRCONFHWYGWFI-UHFFFAOYSA-N
SMILESCCOC(=O)CC(=O)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4200 to 1.4250 (20°C, 589 nm)
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only