Thermo Scientific Chemicals

O-Methylisourea hemisulfate, 94%, Thermo Scientific Chemicals

Catalog number: 233030250
25 g, Each
Thermo Scientific Chemicals

O-Methylisourea hemisulfate, 94%, Thermo Scientific Chemicals

Catalog number: 233030250
25 g, Each
Quantity
Catalog number: 233030250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
52328-05-9
IUPAC Name
methoxymethanimidamide
Molecular Formula
C2H6N2O
InChI Key
RMAHPRNLQIRHIJ-UHFFFAOYSA-N
SMILES
COC(N)=N
Appearance (Color)
White
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder or crystals
Titration with NaOH
>=92 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text