4-Fluoro-2-nitroaniline, 99%
4-Fluoro-2-nitroaniline, 99%
4-Fluoro-2-nitroaniline, 99%
4-Fluoro-2-nitroaniline, 99%
Thermo Scientific Chemicals

4-Fluoro-2-nitroaniline, 99%

CAS: 364-78-3 | C6H5FN2O2 | 156.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
Packaging:
Glass bottle
Catalog number 233070100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS364-78-3
SpecificationsSpecification SheetSpecification Sheet
Melting point91°C to 95°C
GC>=98.5 %
Infrared spectrumConforms
Appearance (Color)Orange to brown
Appearance (Form)Powder or crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link