Thermo Scientific™

Easy-Titer™ Human IgG (H+L) Assay Kit

Catalog number: 23310
Have Questions?
Thermo Scientific™

Easy-Titer™ Human IgG (H+L) Assay Kit

Catalog number: 23310
Have Questions?
Catalog Number
23310
Unit Size
96 tests
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
2331096 tests
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific Easy-Titer Human IgG (H+L) Assay Kit includes antibody-sensitized microspheres to measure the specific concentration of antibodies by an easy and rapid microagglutination technique using standard microplates and UV-Vis plate reader (spectrophotometer). This kit is specific for human IgG and, unlike total protein assays, can specifically measure the concentration of target antibody in samples (e.g., serum, plasma, culture supernatant) that contain other proteins. It is sensitive, requiring very small sample volumes. Antibody concentration is determined from the assay response (absorbance) by comparison to a standard curve prepared using dilutions of a known antibody sample (sold separately).

General features of Easy-Titer Antibody Assay Kits:

Antibody-based specificity—measure concentration of target antibody in a sample, not just total protein; no need to purify antibody to assess its concentration
Faster and easier than ELISA—three-component, homogenous assay; 10 minutes total incubation time
No special equipment needed—uses standard vortex mixer, pipetter, 96-well microplate, plate shaker and reader (measure absorbance at 340nm or 400nm)
Sensitive—assay range (standard curve) 8 to 500 ng/mL; use sample at 15 to 300 ng/mL for optimal results
Reproducible—coefficient of variation < 5%; error depends on dilution and pipetting technique
Antibody standards sold separately—see product list for suggested products; use any antibody standard with proper target identity and known concentration (greater than 10 μg/mL)
Kits for five popular targets—choose a kit specific for a particular species and class of immunoglobulin; no cross-reaction with other species and classes of the target antibody

Easy-Titer Assay Kits detect and measure specific target antibodies using agglutination of microspheres that are coated ('sensitized') with the specific anti-IgG or IgM polyclonal antibodies. In the appropriate aqueous buffer (supplied in kit), the monodispersed antibody-coated microspheres (> 1 μM diameter) have highest absorptivity (λ-max) to incident light having a wavelength (340nm) that is equal to approximately half their diameter. When sample is added, two or more microspheres bind to each antibody target via their coated specific polyclonal antibodies, and this agglutination into effectively larger apparent spheres results in proportional decrease in absorptivity (lower absorbance).

Typical microagglutination assays depend on a change in light-scattering and corresponding change in transmittance, to which absorbance is inversely related. Easy-Titer Assay Kits use a special dilution buffer whose refractive index eliminates the effect of light-scattering on the monodispersed microspheres for the measurement wavelength used. Because of this, the final 10- to 20-fold dilution of the sample for use in the assay must be done using the Dilution Buffer supplied in the kit.

Easy-Titer Antibody Assays are faster and easier than ELISA:
• Prepare standards (5 to 500 ng/mL) by diluting purified antibody in Kit Dilution Buffer.
• Prepare samples by diluting in Dilution Buffer to within assay range (8 to 500 ng/mL).
• Vortex vial of microsphere beads to create homogeneous suspension.
• Pipette 20 μL of bead suspension and 20 μL of each sample and standard into 96-well microplate wells.
• Incubate microplate for 5 minutes with vigorous mixing.
• Add 100 μL of Kit Blocking Reagent.
• Incubate microplate for 5 minutes with vigorous mixing.
• Measure absorbance on standard plate reader (340nm or 405nm).
• Plot standard curve and interpolate samples to determine concentration.

Related Products
Easy-Titer™ Mouse IgG Assay Kit
Easy-Titer™ Rabbit IgG Assay Kit
Easy-Titer™ Human IgG (gamma chain) Assay Kit
Easy-Titer™ Human IgM Assay Kit
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Description
Easy-Titer Human IgG (H + L) Assay Kit
For Use With (Application)
Solution-based Detection, Absorbance
For Use With (Equipment)
Spectrophotometer, Microplate Reader
Quantity
96 Tests
Assay
Protein Quantitation Assay, Isotyping Assay, Immunoassay
Detection Method
Colorimetric
Product Line
Easy-Titer™
Target Specificity
Human IgG

Contents & Storage

Sufficient For: 96 total tests (including 9 standards per use)
• Goat Anti-Human IgG Sensitized Polystyrene Beads, 2mL
• Easy-Titer Dilution Buffer, 30 mL
• Easy-Titer Blocking Buffer, 15 mL

Store at 4°C (do not freeze).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text