Thermo Scientific Chemicals

Benzo[b]furan-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 233260050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Benzo[b]furan-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 233260050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 233260050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
496-41-3
IUPAC Name
1-benzofuran-2-carboxylic acid
Molecular Formula
C9H6O3
InChI Key
OFFSPAZVIVZPHU-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC2=CC=CC=C2O1
Appearance (Color)
White to yellow
Titration with NaOH
>=97.5 %
Infrared spectrum
Conforms
Appearance (Form)
Powder or crystals
Melting point
192°C to 196°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text