Benzo[b]furan-2-carboxylic acid, 98%, Thermo Scientific Chemicals
Benzo[b]furan-2-carboxylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzo[b]furan-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number 233260250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS496-41-3
IUPAC Name1-benzofuran-2-carboxylic acid
Molecular FormulaC9H6O3
InChI KeyOFFSPAZVIVZPHU-UHFFFAOYSA-N
SMILESOC(=O)C1=CC2=CC=CC=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Titration with NaOH>=97.5 %
Infrared spectrumConforms
Appearance (Form)Powder or crystals
Melting point192°C to 196°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only