Thermo Scientific Chemicals

Ethyl (S)-(+)-2-pyrrolidone-5-carboxylate, 99%, Thermo Scientific Chemicals

Catalog number: 233320500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl (S)-(+)-2-pyrrolidone-5-carboxylate, 99%, Thermo Scientific Chemicals

Catalog number: 233320500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 233320500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7149-65-7
IUPAC Name
ethyl 5-oxopyrrolidine-2-carboxylate
Molecular Formula
C7H11NO3
InChI Key
QYJOOVQLTTVTJY-UHFFFAOYNA-N
SMILES
CCOC(=O)C1CCC(=O)N1
Appearance (Color)
White to cream
Infrared spectrum
Conforms
Specific optical rotation
-2° to -5° (20°C, 589 nm) (c=5, H2O)
Appearance (Form)
Low melting solid
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text