Polysorbate 20, Thermo Scientific Chemicals
This product ships via ground from US northeast.
Polysorbate 20, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polysorbate 20, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
5 mL
250 mL
1 L
5 L
Packaging Type:
View all
Plastic drum
Catalog number 233360250
Price (USD)/ Each
840.65
Online exclusive
934.00 
Save 93.35 (10%)
-
Add to cart
Quantity:
25 L
Packaging Type:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
840.65
Online exclusive
934.00 
Save 93.35 (10%)
Add to cart
Polysorbate 20, Thermo Scientific Chemicals
Catalog number233360250
Price (USD)/ Each
840.65
Online exclusive
934.00 
Save 93.35 (10%)
-
Add to cart
Chemical Identifiers
CAS9005-64-5
Molecular Formula(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
InChI KeyHMFKFHLTUCJZJO-UHFFFAOYNA-N
SMILESCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
MDL NumberMFCD00165986
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to slightly hazy yellow to brown-yellow
Appearance (Form)Viscous liquid
AppearanceMay develop some turbidity or precipitate
Infrared spectrumConforms
Water=<3 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only