233PK also known as 233
Full specifications
Pack of 250 5.4 mL 3.1 mL Non-sterile Bulk 250/Pk 8Pk/Cs 2000/Cs - Bulb Draw (Metric) 3.1 mL Description Extended Fine Tip, Large Bulb Transfer Pipette, 5.4mL Diameter (Metric) Outer 3.0 mm Material LDPE Sterility Non-sterile Drops/mL 50 Graduated No Packaging Type Bulk 250/Pk 8Pk/Cs 2000/Cs Volume (Metric) 5.4 mL Length (Metric) 10.4 cm Type Fine Tip Transfer Pipet Disposable Single-use Unit Size Pack of 250
231-1S Full specifications
Case of 3200 1.7 mL 1.0 mL Sterile Individually Wrapped 400/Pk 8Pk/Cs 3200/Cs 781.11 Online price 971.00
- Bulb Draw (Metric) 1.0 mL Description Extended Fine Tip Transfer Pipets, Small Bulb, 1.7mL Diameter (Metric) Outer 3.0 mm Material LDPE Sterility Sterile Drops/mL 50 Graduated No Packaging Type Individually Wrapped 400/Pk 8Pk/Cs 3200/Cs Volume (Metric) 1.7 mL Length (Metric) 10.4 cm Type Fine Tip Transfer Pipet Disposable Single-use Unit Size Case of 3200
232-11 Full specifications
Case of 5000 5.8 mL 3.3 mL Non-sterile Bulk 500/Pk 10Pk/Cs 5000/Cs - Bulb Draw (Metric) 3.3 mL Description Fine Tip Standard Bulb Transfer Pipette, 5.8mL Diameter (Metric) Outer 4.6 mm Material LDPE Sterility Non-sterile Drops/mL 53 Graduated No Packaging Type Bulk 500/Pk 10Pk/Cs 5000/Cs Volume (Metric) 5.8 mL Length (Metric) 14.7 cm Type Fine Tip Transfer Pipet Disposable Single-use Unit Size Case of 5000
231-20S Full specifications
Case of 4000 1.7 mL 1.0 mL Sterile 20/Bag, 500/Pk., 4000/Cs. - Bulb Draw (Metric) 1.0 mL Description Extended Fine Tip Transfer Pipets, Small Bulb, 1.7mL Diameter (Metric) Outer 3.0 mm Material LDPE Sterility Sterile Drops/mL 50 Graduated No Packaging Type 20/Bag, 500/Pk., 4000/Cs. Volume (Metric) 1.7 mL Length (Metric) 10.4 cm Type Fine Tip Transfer Pipet Disposable Single-use Unit Size Case of 4000
231 Full specifications
Case of 3200 1.7 mL 1.0 mL Non-sterile Bulk 400/Pk 8Pk/Cs 3200/Cs 321.77 Online price 523.00
- Bulb Draw (Metric) 1.0 mL Description Extended Fine Tip Transfer Pipets, Small Bulb, 1.7mL Diameter (Metric) Outer 3.0 mm Material LDPE Sterility Non-sterile Drops/mL 50 Graduated No Packaging Type Bulk 400/Pk 8Pk/Cs 3200/Cs Volume (Metric) 1.7 mL Length (Metric) 10.4 cm Type Fine Tip Transfer Pipet Disposable Single-use Unit Size Case of 3200
231PK also known as 231
Full specifications
Pack of 400 1.7 mL 1.0 mL Non-sterile Bulk 400/Pk 8Pk/Cs 3200/Cs - Bulb Draw (Metric) 1.0 mL Description Extended Fine Tip Transfer Pipets, Small Bulb, 1.7mL Diameter (Metric) Outer 3.0 mm Material LDPE Sterility Non-sterile Drops/mL 50 Graduated No Packaging Type Bulk 400/Pk 8Pk/Cs 3200/Cs Volume (Metric) 1.7 mL Length (Metric) 10.4 cm Type Fine Tip Transfer Pipet Disposable Single-use Unit Size Pack of 400
233 Full specifications
Case of 2000 5.4 mL 3.1 mL Non-sterile Bulk 250/Pk 8Pk/Cs 2000/Cs - Bulb Draw (Metric) 3.1 mL Description Extended Fine Tip, Large Bulb Transfer Pipette, 5.4mL Diameter (Metric) Outer 3.0 mm Material LDPE Sterility Non-sterile Drops/mL 50 Graduated No Packaging Type Bulk 250/Pk 8Pk/Cs 2000/Cs Volume (Metric) 5.4 mL Length (Metric) 10.4 cm Type Fine Tip Transfer Pipet Disposable Single-use Unit Size Case of 2000
233-1S Full specifications
Case of 3200 5.4 mL 3.1 mL Sterile Individually Wrapped 400/Pk 8Pk/Cs 3200/Cs 378.00 Online price 1,199.00
- Bulb Draw (Metric) 3.1 mL Description Extended Fine Tip, Large Bulb Transfer Pipette, 5.4mL Diameter (Metric) Outer 3.0 mm Material LDPE Sterility Sterile Drops/mL 50 Graduated No Packaging Type Individually Wrapped 400/Pk 8Pk/Cs 3200/Cs Volume (Metric) 5.4 mL Length (Metric) 10.4 cm Type Fine Tip Transfer Pipet Disposable Single-use Unit Size Case of 3200
233-20S Full specifications
Case of 4000 5.4 mL 3.1 mL Sterile 20/Bag, 400/Pk., 4000/Cs. 781.11 Online price 971.00
- Bulb Draw (Metric) 3.1 mL Description Extended Fine Tip, Large Bulb Transfer Pipette, 5.4mL Diameter (Metric) Outer 3.0 mm Material LDPE Sterility Sterile Drops/mL 50 Graduated No Packaging Type 20/Bag, 400/Pk., 4000/Cs. Volume (Metric) 5.4 mL Length (Metric) 10.4 cm Type Fine Tip Transfer Pipet Disposable Single-use Unit Size Case of 4000
232-1S Full specifications
Case of 3200 5.8 mL 3.3 mL Sterile Individually Wrapped 400/Pk 8Pk/Cs 3200/Cs - Bulb Draw (Metric) 3.3 mL Description Fine Tip Standard Bulb Transfer Pipette, 5.8mL Diameter (Metric) Outer 4.6 mm Material LDPE Sterility Sterile Drops/mL 53 Graduated No Packaging Type Individually Wrapped 400/Pk 8Pk/Cs 3200/Cs Volume (Metric) 5.8 mL Length (Metric) 14.7 cm Type Fine Tip Transfer Pipet Disposable Single-use Unit Size Case of 3200