Thermo Scientific™

Pierce™ Controlled Protein-Protein Crosslinking Kit

Catalog number: 23456
Have Questions?
Thermo Scientific™

Pierce™ Controlled Protein-Protein Crosslinking Kit

Catalog number: 23456
Have Questions?
Catalog Number
23456
Unit Size
1 reaction
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
234561 reaction
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific Pierce Controlled Protein-Protein Crosslinking Kit contains essential crosslinkers, modification reagents and buffers for conjugating antibodies, enzymes and other purified proteins needed as probes.

This crosslinking kit contains everything you need to help you insert an unbreakable molecular bridge between practically any two purified proteins and do it successfully right out of the box.

Features of the Controlled Protein-Protein Crosslinking Kit:

• Easy-to-control, stepwise coupling of two proteins—You are in charge of the chemistry. Stop and restart at convenient points or go to completion. It is your choice.
• Better protocol…logically presented and clearly worded instructions which will be appreciated by novice and expert—Instructions teach the method and chemistry of this crosslinking strategy to the user while providing a high likelihood of success. Great for life scientists attempting to prepare a conjugate or crosslink two proteins for the first time.
• Reliable non-cleavable heterobifunctional crosslinking agent—A very popular choice for a crosslinking agent was selected for the kit offering proven chemistry ,highly stabile intermediates and efficient formation of the target conjugate adding to the ease of use and likelihood of success.
• All reagents necessary to affect the crosslinking—Cuts costs over purchasing 'catalog size' packaging of all the components by more than 60%. If it's not in the kit, you don't need it!
• Includes disulfide (S-S) reductants and thiolation reagents—Allows for all possibilities regarding the state of the—SH group on one of the proteins to be coupled.
• Ellman's Reagent colorimetric—SH group determination chemistry built into the protocols—You have the option to see how well your reactions went. eg. determine the extent of—SH introduction or estimate the incorporation of maleimide reactive group.

Related Products
Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate)
WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Reactivity
Amine-Sulfhydryl
Cleavable
No
Description
Controlled Protein-Protein Crosslinking Kit
Molecular Weight (g/mol)
436.37
PEGylated
No
Spacer Arm Length
11.6 Å
Cell Permeability
No
Shipping Condition
Ambient
Product Line
Pierce™
Labeling Method
Chemical Labeling
Crosslinker Type
Heterobifunctional
Reactive Moiety
Maleimide, Sulfo-NHS Ester
Spacer
Short (<10 Å)
Form
Kit
Quantity
1 Reaction
Solubility
Water
Format
Kit
Water Soluble
Yes
Product Type
Crosslinker

Contents & Storage

Sufficient For: One conjugation with two purified proteins (up to 5 mg each)
• Sulfo-SMCC Crosslinking Agent, 2 mg
• Activation/Conjugation Buffer (10X), 20 mL
• PBS packs (each makes 500mL), 2 packs
• 2-Mercaptoethylamine-HCl, 6 mg
• Immobilized-Reductant, 0.2 mL
• SATA, 2 mg
• Dimethylformamide (DMF), 1 mL
• Hydroxylamine-HCl, 5 mg
• Dextran Desalting Column, 2 x 10 mL
• Column Extender, 2 ea.
• Cysteine-HCl, 20 mg
• Ellman's Reagent, 2 mg

Store at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text