Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Volume (Metric) | Sterility | Bulb Draw (Metric) | Packaging Type | Unit Size |
---|---|---|---|---|---|
235-1S | 5.8 mL | Sterile | 3.7 mL | Individually Wrapped | Case of 3200 |
231-1S | 1.7 mL | Sterile | 1.0 mL | Individually Wrapped | Case of 3200 |
232-11 | 5.8 mL | Non-sterile | 3.3 mL | Bulk 500/Pk 10Pk/Cs 5000/Cs | Case of 5000 |
231-20S | 1.7 mL | Sterile | 1.0 mL | 20/Bag, 500/Pk., 4000/Cs. | Case of 4000 |
231 | 1.7 mL | Non-sterile | 1.0 mL | Bulk 400/Pk 8Pk/Cs 3200/Cs | Case of 3200 |
231PK also known as 231 | 1.7 mL | Non-sterile | 1.0 mL | Bulk 400/Pk 8Pk/Cs 3200/Cs | Pack of 400 |
233 | 5.4 mL | Non-sterile | 3.1 mL | Bulk 250/Pk 8Pk/Cs 2000/Cs | Case of 2000 |
233PK also known as 233 | 5.4 mL | Non-sterile | 3.1 mL | Bulk 250/Pk 8Pk/Cs 2000/Cs | Pack of 250 |
233-1S | 5.4 mL | Sterile | 3.1 mL | Individually Wrapped | Case of 3200 |
233-20S | 5.4 mL | Sterile | 3.1 mL | 20/Bag, 400/Pk., 4000/Cs. | Case of 4000 |
232-1S | 5.8 mL | Sterile | 3.3 mL | Individually Wrapped | Case of 3200 |
232-1SPK also known as 232-1S | 5.8 mL | Sterile | 3.3 mL | Individually Wrapped | Pack of 400 |
235-20S | 5.8 mL | Sterile | 3.7 mL | 20/Bag, 500/Pk., 4000/Cs. | Case of 4000 |
232-20S | 5.8 mL | Sterile | 3.3 mL | 20/Bag, 500/Pk., 4000/Cs. | Case of 4000 |
235 | 5.8 mL | Non-sterile | 3.7 mL | Bulk 500/Pk 10Pk/Cs 5000/Cs | Case of 5000 |
235PK also known as 235 | 5.8 mL | Non-sterile | 3.7 mL | Bulk 500/Pk 10Pk/Cs 5000/Cs | Pack of 500 |
234 | 8.9 mL | Non-sterile | 4.8 mL | Bulk 400/Pk 10Pk/Cs 4000/Cs | Case of 4000 |
234PK also known as 234 | 8.9 mL | Non-sterile | 4.8 mL | Bulk 400/Pk 10Pk/Cs 4000/Cs | Pack of 400 |
234-1S | 8.9 mL | Sterile | 4.8 mL | Individually Wrapped | Case of 3200 |
234-1SPK also known as 234-1S | 8.9 mL | Sterile | 4.8 mL | Individually Wrapped | Pack of 400 |
234-20S | 8.9 mL | Sterile | 4.8 mL | 20/Bag, 400/Pk., 3200/Cs. | Case of 3200 |