Butylated hydroxyanisole, 96%, Thermo Scientific Chemicals
Butylated hydroxyanisole, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Butylated hydroxyanisole, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Plastic bottle
Catalog number 235230025
Price (USD)/ Each
307.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
362.00 
Save 54.35 (15%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
307.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
362.00 
Save 54.35 (15%)
Add to cart
Butylated hydroxyanisole, 96%, Thermo Scientific Chemicals
Catalog number235230025
Price (USD)/ Each
307.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
362.00 
Save 54.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2-tert-butyl-4-methoxyphenol
Molecular FormulaC11H16O2
InChI KeyMRBKEAMVRSLQPH-UHFFFAOYSA-N
SMILESCOC1=CC=C(O)C(=C1)C(C)(C)C
Molecular Weight (g/mol)180.25
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals or flakes
Appearance (Color)White to light yellow
GC>=95 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only