Butylated hydroxyanisole, 96%
Butylated hydroxyanisole, 96%
Butylated hydroxyanisole, 96%
Butylated hydroxyanisole, 96%
Thermo Scientific Chemicals

Butylated hydroxyanisole, 96%

CAS: 25013-16-5 | C11H16O2 | 180.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 235235000
Price (USD)
93.65
Special offer
Online exclusive
Ends: 26-Sep-2025
116.00
Save 22.35 (19%)
Each
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
93.65
Special offer
Online exclusive
Ends: 26-Sep-2025
116.00
Save 22.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS25013-16-5
IUPAC Name2-tert-butyl-4-methoxyphenol
Molecular FormulaC11H16O2
InChI KeyMRBKEAMVRSLQPH-UHFFFAOYSA-N
SMILESCOC1=CC=C(O)C(=C1)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
GC>=95 %
Appearance (Form)Crystals or flakes
Infrared spectrumConforms
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0464174Certificate of AnalysisJul 02, 2025235230025, 235235000, 235231000
A0468529Certificate of AnalysisMay 17, 2025235230025, 235235000, 235231000
A0464174Certificate of AnalysisJul 03, 2024235230025, 235235000, 235231000
A0444369Certificate of AnalysisAug 25, 2023235230025, 235235000, 235231000
A0399813Certificate of AnalysisDec 16, 2022235230025, 235235000, 235231000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection