Ethyl 2-(bromomethyl)acrylate, 97%, Thermo Scientific Chemicals
Ethyl 2-(bromomethyl)acrylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 2-(bromomethyl)acrylate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 235360010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS17435-72-2
IUPAC Nameethyl 2-(bromomethyl)prop-2-enoate
Molecular FormulaC6H9BrO2
InChI KeyMTCMFVTVXAOHNQ-UHFFFAOYSA-N
SMILESCCOC(=O)C(=C)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=96.0 %
Refractive index1.4770 to 1.4810 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only