Inositol hexaphosphoric acid, 40-50 wt.% aqueous solution
Inositol hexaphosphoric acid, 40-50 wt.% aqueous solution
Thermo Scientific Chemicals

Inositol hexaphosphoric acid, 40-50 wt.% aqueous solution

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 235372500
Price (USD)/ Each
61.65
Online exclusive
72.30 
Save 10.65 (15%)
-
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
61.65
Online exclusive
72.30 
Save 10.65 (15%)
Add to cart
Inositol hexaphosphoric acid, 40-50 wt.% aqueous solution
Catalog number235372500
Price (USD)/ Each
61.65
Online exclusive
72.30 
Save 10.65 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name{[(1R,2R,3S,4R,5s,6S)-2,3,4,5,6-pentakis(phosphonooxy)cyclohexyl]oxy}phosphonic acid
Molecular FormulaC6H18O24P6
InChI KeyIMQLKJBTEOYOSI-GPIVLXJGSA-N
SMILESOP(O)(=O)O[C@H]1[C@H](OP(O)(O)=O)[C@@H](OP(O)(O)=O)[C@H](OP(O)(O)=O)[C@H](OP(O)(O)=O)[C@@H]1OP(O)(O)=O
Molecular Weight (g/mol)660.03
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Titration with NaOH45 to 55 % w/w
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only