Dimethyl-β-cyclodextrin, methylated β-cyclodextrins, 96%, Thermo Scientific Chemicals
Dimethyl-β-cyclodextrin, methylated β-cyclodextrins, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethyl-β-cyclodextrin, methylated β-cyclodextrins, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 235530010
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
IUPAC Name37,39,41,43,45,47,49-heptamethoxy-5,10,15,20,25,30,35-heptakis(methoxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,38,40,42,44,46,48-heptol
Molecular FormulaC56H98O35
InChI KeyQGKBSGBYSPTPKJ-UHFFFAOYNA-N
SMILESCOCC1OC2OC3C(COC)OC(OC4C(COC)OC(OC5C(COC)OC(OC6C(COC)OC(OC7C(COC)OC(OC8C(COC)OC(OC1C(O)C2OC)C(OC)C8O)C(OC)C7O)C(OC)C6O)C(OC)C5O)C(OC)C4O)C(OC)C3O
Molecular Weight (g/mol)1331.37
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
Appearance (Form)Crystalline powder
TLC=<0.5 % unmodified beta-cyclodextrin
HPLC35.0 to 55.0 % isomer purity
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only