Thermo Scientific Chemicals

Dimethyl-β-cyclodextrin, methylated β-cyclodextrins, 96%, Thermo Scientific Chemicals

Catalog number: 235530050
5 g, Each
Thermo Scientific Chemicals

Dimethyl-β-cyclodextrin, methylated β-cyclodextrins, 96%, Thermo Scientific Chemicals

Catalog number: 235530050
5 g, Each
Quantity
Catalog number: 235530050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
51166-71-3
IUPAC Name
37,39,41,43,45,47,49-heptamethoxy-5,10,15,20,25,30,35-heptakis(methoxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,38,40,42,44,46,48-heptol
Molecular Formula
C56H98O35
InChI Key
QGKBSGBYSPTPKJ-UHFFFAOYNA-N
SMILES
COCC1OC2OC3C(COC)OC(OC4C(COC)OC(OC5C(COC)OC(OC6C(COC)OC(OC7C(COC)OC(OC8C(COC)OC(OC1C(O)C2OC)C(OC)C8O)C(OC)C7O)C(OC)C6O)C(OC)C5O)C(OC)C4O)C(OC)C3O
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
TLC
=<0.5 % unmodified beta-cyclodextrin
HPLC
35.0 to 55.0 % isomer purity

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text