2,6-Diaminoheptanedioic acid, 95%, Thermo Scientific Chemicals
2,6-Diaminoheptanedioic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Diaminoheptanedioic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 235540010
Price (USD)/ Each
45.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
53.40 
Save 7.75 (15%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
45.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
53.40 
Save 7.75 (15%)
Add to cart
2,6-Diaminoheptanedioic acid, 95%, Thermo Scientific Chemicals
Catalog number235540010
Price (USD)/ Each
45.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
53.40 
Save 7.75 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name(2R,6S)-2,6-diazaniumylheptanedioate
Molecular FormulaC7H14N2O4
InChI KeyGMKMEZVLHJARHF-SYDPRGILSA-N
SMILES[NH3+][C@@H](CCC[C@@H]([NH3+])C([O-])=O)C([O-])=O
Molecular Weight (g/mol)190.20
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to beige to tan
Titration with HClO494.0 to 106.0 %
Appearance (Form)Powder and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only