Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers
Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers
Thermo Scientific™
Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers
Secure vessels using these universal platform clamps for orbital shakers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)
236013250/300mL Erlenmeyer Flask ClampMaxQ™ 8000 Shaker
23601025mL Erlenmeyer Flask ClampMaxQ™ 8000 Shaker
23601150mL Erlenmeyer Flask ClampMaxQ™ 8000 Shaker
236012125mL Erlenmeyer Flask ClampMaxQ™ 8000 Shaker
236014500mL Erlenmeyer Flask ClampMaxQ™ 8000 Shaker
2360151L Erlenmeyer Flask ClampMaxQ™ 8000 Shaker
2360162L Erlenmeyer Flask ClampMaxQ™ 8000 Shaker
2360192800mL Fernbach Flask ClampMaxQ™ 8000 Shaker
236025Microplate ClampMaxQ HP 416, 420, 430, 435, 436, 480, 481, 8000
161033Vial/Tube InsertsMaxQ HP 416, 420, 430, 435, 436, 480, 481, 8000
236099Flask Clamp Starter Kit for MaxQ High Performance ShakersMaxQ HP 416, 420, 430, 435, 436, 480, 481, 8000
Catalog number 236013
Price (USD)
45.09
Online Exclusive
50.10
Save 5.01 (10%)
Each
Add to cart
Description:
250/300mL Erlenmeyer Flask Clamp
For Use With (Equipment):
MaxQ™ 8000 Shaker
Price (USD)
45.09
Online Exclusive
50.10
Save 5.01 (10%)
Each
Add to cart
Secure vessels using Thermo Scientific™ Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers. With exceptional innovation, reliability and accuracy, these orbital shakers, platforms and accessories are the optimal choice for a busy lab.
Flask Clamp Starter Kit includes:
  • (1) microplate clamp
  • (2) 25mL flask clamps
  • (2) 50mL flask clamps
  • (2) 125mL flask clamps
  • (4) 250/300mL flask clamps
  • (4) 500mL flask clamps
  • (2) 1L flask clamps
  • (2) 2L flask clamps
  • Replacement screws
Note:
Replacement screws also available separately.
Specifications
Description250/300mL Erlenmeyer Flask Clamp
For Use With (Equipment)MaxQ™ 8000 Shaker
Holds250/300 mL
Product TypeFlask Clamp
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link