EBSS, calcium, magnesium, phenol red
Save up to 29% on your order of cell culture products. View promotion details >
Product Image
Gibco™

EBSS, calcium, magnesium, phenol red

Earle's Balanced Salt Solution (EBSS) is used for a variety of cell culture applications, such as washing cells before dissociation,Read more
Promo ImagePromotion
Promo code:RCELL25
Save up to 29% across the cell culture workflow - research begins with cells
From now until Dec 31, get up to 29% off media, sera, buffers, reagents and other cell culture products to advance your researchLearn More
Have Questions?
Catalog NumberQuantity
24010043Promo Image500 mL
Catalog number 24010043
Price (USD)
24.65
Online Exclusive
26.66
Save 2.01 (8%)
Each
In stock
Add to cart
Quantity:
500 mL
Customize this product
Price (USD)
24.65
Online Exclusive
26.66
Save 2.01 (8%)
Each
Add to cart

Earle's Balanced Salt Solution (EBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation.


This EBSS is modified as follows:
With
• Glucose
• Sodium Bicarbonate
• Calcium
• Magnesium
• Phenol Red


The complete formulation is available.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

Specifications
Concentration1 X
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality270 - 310 mOsm/kg
Product LineGibco™
Product TypeEarle's Balanced Salt Solution (EBSS)
Quantity500 mL
Shelf Life36 Months
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free
FormLiquid
SterilitySterile-filtered
With AdditivesPhenol Red, Glucose, Sodium Bicarbonate, Calcium, Magnesium
Without AdditivesNo Sodium Pyruvate
pH7 to 7.4
Unit SizeEach
Contents & Storage
Storage conditions: 15-30° C.
Shipping conditions: Ambient.
Shelf life: 36 months from date of manufacture.
four-plex-image-COS7-live-cell promo pod

Follow the 5 steps to publication-quality fixed-cell imaging.
Download your free 5-step guide today ›

Frequently asked questions (FAQs)

Do you offer media without phenol red (phenol red-free media)?

Yes, please visit this page for a list of our phenol red-free media (http://tools.thermofisher.com/content/sfs/brochures/Phenol_RedFree_Media_Glance.pdf).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Is it true that phenol red has estrogenic properties?

Phenol red in tissue culture is a weak estrogen. Implications concerning study of estrogen-responsive cells in culture are discussed in Proc Natl Acad Sci U S A 83:2496 (1986).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link