Silica gel, for chromatography, 0.060-0.200 mm, 60 A
Silica gel, for chromatography, 0.060-0.200 mm, 60 A
Silica gel, for chromatography, 0.060-0.200 mm, 60 A
Silica gel, for chromatography, 0.060-0.200 mm, 60 A
Silica gel, for chromatography, 0.060-0.200 mm, 60 A
Thermo Scientific Chemicals

Silica gel, for chromatography, 0.060-0.200 mm, 60 A

For column chromatography. | CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
30 kg
Catalog number 240370300
Price (USD)
1,727.65
Online Exclusive
1,919.00
Save 191.35 (10%)
Each
-
Add to cart
Quantity:
30 kg
Request bulk or custom format
Price (USD)
1,727.65
Online Exclusive
1,919.00
Save 191.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
pH6.5 to 7.5 (5 % suspension in water)
Particle size=<5 % (Undersize)
Residual water2 to 4 % (160°C)
Appearance (Color)White
Specific gravity0.46 to 0.55 kg/l
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link