Thermo Scientific Chemicals

Silica gel, for chromatography, 0.200-0.500mm, 60Å

Catalog number: 240380050
Plastic Drum, 5 kg, Each
Thermo Scientific Chemicals

Silica gel, for chromatography, 0.200-0.500mm, 60Å

Catalog number: 240380050
Plastic Drum, 5 kg, Each
Packaging
Quantity
Catalog number: 240380050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7631-86-9
IUPAC Name
silanedione
Molecular Formula
O2Si
InChI Key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES
O=[Si]=O
Appearance (Color)
White
Specific surface area
480 to 550 m²/g
pH
6.0 to 7.5 (5 % suspension in water)
Residual water
=<4 % (150°C, 2 h)
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications:

For column chromatography

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text