Potassium methoxide, pure, 0.1 N solution in Toluene/Methanol, Thermo Scientific Chemicals
Potassium methoxide, pure, 0.1 N solution in Toluene/Methanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium methoxide, pure, 0.1 N solution in Toluene/Methanol, Thermo Scientific Chemicals

Have Questions?
Quantity:
500 mL
Packaging:
Glass bottle
Catalog number 240605000
Price (USD)/ Each
197.65
Online exclusive
219.00 
Save 21.35 (10%)
-
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
197.65
Online exclusive
219.00 
Save 21.35 (10%)
Add to cart
Potassium methoxide, pure, 0.1 N solution in Toluene/Methanol, Thermo Scientific Chemicals
Catalog number240605000
Price (USD)/ Each
197.65
Online exclusive
219.00 
Save 21.35 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Namepotassium methanolate
Molecular FormulaCH3KO
InChI KeyBDAWXSQJJCIFIK-UHFFFAOYSA-N
SMILES[K+].C[O-]
Molecular Weight (g/mol)70.13
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Clear colorless
Titration with HCl0.095 to 0.105 N
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only