Search Thermo Fisher Scientific
Product Type | Multiskan Verification Plate, 16 wavelengths |
For Use With (Equipment) | Multiskan FC |
Plate Type | Verification Plate |
Product Line | Multiskan™ |
Includes | NPL (National Physical Laboratory) traceable Multiskan™ Verification Plate Certificate of Calibration USB memory device including SkanIt™ software sessions and reference files Carrying case for Multiskan™ Verification Plate |
Unit Size | Each |
Product Type | Multiskan Verification Plate, 8 wavelengths |
For Use With (Equipment) | Multiskan FC |
Plate Type | Verification Plate |
Product Line | Multiskan™ |
Includes | NPL (National Physical Laboratory) traceable Multiskan™ Verification Plate Certificate of Calibration USB memory device including SkanIt™ software sessions and reference files Carrying case for Multiskan™ Verification Plate |
Unit Size | Each |
Product Type | Verification Plate |
For Use With (Equipment) | Varioskan LUX Microplate Reader, Luminoskan |
Plate Type | Verification Plate |
Includes | Lumiwell Luminometric Verification Plate Certificate of Calibration Carrying case Lumiwell Verification Plate |
Unit Size | Each |
Product Type | Spectrophotometric Verification Plate |
For Use With (Equipment) | Varioskan LUX Microplate Reader, Multiskan Sky, Multiskan Skyhigh |
Plate Type | Verification Plate |
Product Line | Varioskan™ |
Includes | NPL (UK National Physical Laboratory) and NIST (US National Institute for Standards and Technology) traceable Spectrophotometric Verification Plate Certificate of Calibration Adapter for Spectrophotometric Verification Plate USB memory device including SkanIt™ software sessions and reference files Carrying case for Spectrophotometric Verification Plate |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Product Type | Includes | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
24072805 | Each | Multiskan Verification Plate, 16 wavelengths | NPL (National Physical Laboratory) traceable Multiskan™ Verification Plate Certificate of Calibration USB memory device including SkanIt™ software sessions and reference files Carrying case for Multiskan™ Verification Plate | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
24072800 | Each | Multiskan Verification Plate, 8 wavelengths | NPL (National Physical Laboratory) traceable Multiskan™ Verification Plate Certificate of Calibration USB memory device including SkanIt™ software sessions and reference files Carrying case for Multiskan™ Verification Plate | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2806460 | Each | Verification Plate | Lumiwell Luminometric Verification Plate Certificate of Calibration Carrying case Lumiwell Verification Plate | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
N03394 | Each | Spectrophotometric Verification Plate | NPL (UK National Physical Laboratory) and NIST (US National Institute for Standards and Technology) traceable Spectrophotometric Verification Plate Certificate of Calibration Adapter for Spectrophotometric Verification Plate USB memory device including SkanIt™ software sessions and reference files Carrying case for Spectrophotometric Verification Plate | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|