Search Thermo Fisher Scientific
Catalog Number | Description |
---|---|
24075410 | KingFisher Flex 96 PCR heating block |
24074410 | KingFisher Flex 96 PCR Head |
24074411 | KingFisher Flex 96 PCR head and heating block |
24074431 | KingFisher Flex 96 Deep Well head and deep well and KF heating blocks |
24074420 | KingFisher Flex 96 KF head |
24074421 | KingFisher Flex 96 KF head and heating block |
24074441 | KingFisher Flex 24 Deep Well head and heating block |
5400620 | KingFisher 96 Plate |
A39952 | KingFisher 96 Plate, Extended Warranty Package |
A39966 | KingFisher 96 Plate with SmartStart |
Find a flexible solution for different types of sample processing needs. With 24 configurations, customers can raise the processing volume up to 5mL, resulting in increased yield of the purified product. For higher throughput needs, 96 samples can be processed with 96 magnet head.
• Fully automated system yields high-speed purification of nucleic acids, proteins and cells.
• High throughput system processes up to 96 samples in less than 20 minutes.
• Expands sample volume by 10-fold with the 24-well format.
• Open and flexible system lets the customer use anymagnetic particle-based kit tomeet the application demands.
• Easy-to-use Thermo Scientific™ BindIt™ Software provides instrument control, protocol creation andmodification.
• Ready-made protocols for different type of applications are available.
Recommended for:
• Genomics and proteomics
• Target identification
• Veterinary assays
• Biomarker discovery
• Quality control
• Forensics