Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

KingFisher™ Flex 96 heating block

Catalog number: 24075420
Have Questions?
Thermo Scientific™

KingFisher™ Flex 96 heating block

Catalog number: 24075420
Have Questions?
Catalog Number
24075420
Unit Size
Each
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
24075420Each
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The KingFisher™ Flex 96 KF Heating Block is compatible with the KingFisher™ 96 KF Microplate (200 μL) and the MagMAX™ Express-96 Standard Plate when used on the KingFisher™ Flex Magnetic Particle Processor with 96 Deep-Well Head. This heating block is also compatible with the KingFisher™ Flex Microtiter Deep Well 96 Plate and the MagMAX™ Express-96 Deep Well plates.

Specifications

Format
96-well Plate
Capacity
96 Samples
For Use With (Equipment)
KingFisher™ Flex
Product Line
KingFisher™
Shipping Condition
Room Temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text