Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

KingFisher™ Flex Deep Well heating block

Catalog number: 24075430
Have Questions?
Thermo Scientific™

KingFisher™ Flex Deep Well heating block

Catalog number: 24075430
Have Questions?
Catalog Number
24075430
Unit Size
Each
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
24075430Each
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The KingFisher Flex 96 Deep-Well Heating Block is compatible with the KingFisher Flex Microtiter Deep-Well 96 Plate and the MagMAX Express-96 Deep-Well Plate when used on the KingFisher Flex Magnetic Particle Processor with 96 Deep-Well Head. Please note that the KingFisher Flex 96 Deep-Well Heating Block is not compatible with the KingFisher 96 microplate (200 μL).

Specifications

Format
96 Deep-well Plate
Capacity
96 Samples
For Use With (Equipment)
KingFisher™ Flex
Product Line
KingFisher™
Shipping Condition
Room Temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text