1x1 image pixel for data collection
Magnesium stearate, 3.8-5.0% Mg
Magnesium stearate, 3.8-5.0% Mg
Magnesium stearate, 3.8-5.0% Mg
Magnesium stearate, 3.8-5.0% Mg
Thermo Scientific Chemicals

Magnesium stearate, 3.8-5.0% Mg

CAS: 557-04-0 | C36H70MgO4 | 591.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 240760010
Price (USD)
53.70
Each
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
53.70
Each
Add to cart
Chemical Identifiers
CAS557-04-0
IUPAC Namemagnesium(2+) dioctadecanoate
Molecular FormulaC36H70MgO4
InChI KeyHQKMJHAJHXVSDF-UHFFFAOYSA-L
SMILES[Mg++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=40.0 % relative content of stearic acid
Heavy metals (as Pb)=<40 ppm
Chloride (Cl)=<1000 ppm
Appearance (Color)White to almost white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection