Thermo Scientific Chemicals

Methyl 4-chloroacetoacetate, 97%, Thermo Scientific Chemicals

Catalog number: 241620250
25 mL, Each
Thermo Scientific Chemicals

Methyl 4-chloroacetoacetate, 97%, Thermo Scientific Chemicals

Catalog number: 241620250
25 mL, Each
Quantity
Catalog number: 241620250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
32807-28-6
IUPAC Name
methyl 4-chloro-3-oxobutanoate
Molecular Formula
C5H7ClO3
InChI Key
HFLMYYLFSNEOOT-UHFFFAOYSA-N
SMILES
COC(=O)CC(=O)CCl
Refractive index
1.4560 to 1.4580 (20°C, 589 nm)
Appearance (Color)
Clear light yellow to light orange
Infrared spectrum
Conforms
Appearance (Form)
Liquid
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text