Thermo Scientific Chemicals

Silica gel, for chromatography, 0.035-0.070 mm, 90 A, Thermo Scientific Chemicals

Catalog number: 241650010
1 kg, Plastic Bottle, Each
Thermo Scientific Chemicals

Silica gel, for chromatography, 0.035-0.070 mm, 90 A, Thermo Scientific Chemicals

Catalog number: 241650010
1 kg, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 241650010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7631-86-9
IUPAC Name
silanedione
Molecular Formula
O2Si
InChI Key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES
O=[Si]=O
Bulk density
385 to 440 g/l
Residual water
2 to 4 % (160°C)
Particle size
=<10 % undersized particles
Appearance (Form)
Powder
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For column chromatography
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text