Silica gel, for column chromatography, 0.060-0.200mm, 40Å
Silica gel, for column chromatography, 0.060-0.200mm, 40Å
Thermo Scientific Chemicals

Silica gel, for column chromatography, 0.060-0.200mm, 40Å

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Catalog number 241660050
Price (USD)/ Each
444.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
523.00 
Save 78.35 (15%)
-
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (USD)/ Each
444.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
523.00 
Save 78.35 (15%)
Add to cart
Silica gel, for column chromatography, 0.060-0.200mm, 40Å
Catalog number241660050
Price (USD)/ Each
444.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
523.00 
Save 78.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Residual water2 to 4 % (160°C)
Particle size=<10 % (Undersize)
Appearance (Form)Powder
pH5.5 to 8.0 (5 % suspension in water)
Particle size0.060 to 0.200 mm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For column chromatography

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only