5-Fluoro-2-nitrophenol, 99%, Thermo Scientific Chemicals
5-Fluoro-2-nitrophenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Fluoro-2-nitrophenol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 241970250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name5-fluoro-2-nitrophenol
Molecular FormulaC6H4FNO3
InChI KeyQQURWFRNETXFTN-UHFFFAOYSA-N
SMILESOC1=CC(F)=CC=C1[N+]([O-])=O
Molecular Weight (g/mol)157.10
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder or crystals or crystalline mass and/or
GC>=98.5 %
Appearance (Form)chunks
Melting point32°C to 35°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only