242-1SPK also known as 242-1S
Full specifications
Pack of 500 4.3 mL 3.1 mL Sterile Individually Wrapped 500/Pk 8Pk/Cs 4000/Cs - Bulb Draw (Metric) 3.1 mL Description Transfer Pipette, SEDI-PET, Short Stem, Long Bulb, 4.3mL Diameter (Metric) Outer 2.5 mm Material LDPE Sterility Sterile Drops/mL 24 Graduated No Packaging Type Individually Wrapped 500/Pk 8Pk/Cs 4000/Cs Volume (Metric) 4.3 mL Length (Metric) 8.5 cm Type Narrow Stem Transfer Pipet Disposable Single-use Unit Size Pack of 500
241 Full specifications
Case of 4000 1.3 mL 0.8 mL Non-sterile Bulk 500/Pk 8Pk/Cs 4000/Cs - Bulb Draw (Metric) 0.8 mL Description Transfer Pipette, SEDI-PET, Short Stem, 1.3mL Diameter (Metric) Outer 2.5 mm Material LDPE Sterility Non-sterile Drops/mL 26 Graduated No Packaging Type Bulk 500/Pk 8Pk/Cs 4000/Cs Volume (Metric) 1.3 mL Length (Metric) 6.4 cm Type Narrow Stem Transfer Pipet Disposable Single-use Unit Size Case of 4000
241PK also known as 241
Full specifications
Pack of 500 1.3 mL 0.8 mL Non-sterile Bulk 500/Pk 8Pk/Cs 4000/Cs - Bulb Draw (Metric) 0.8 mL Description Transfer Pipette, SEDI-PET, Short Stem, 1.3mL Diameter (Metric) Outer 2.5 mm Material LDPE Sterility Non-sterile Drops/mL 26 Graduated No Packaging Type Bulk 500/Pk 8Pk/Cs 4000/Cs Volume (Metric) 1.3 mL Length (Metric) 6.4 cm Type Narrow Stem Transfer Pipet Disposable Single-use Unit Size Pack of 500
241-20S Full specifications
Case of 4000 1.3 mL 0.8 mL Sterile 20/Bag, 500/Pk., 4000/Cs. Request A Quote - - Bulb Draw (Metric) 0.8 mL Description Transfer Pipette, SEDI-PET, Short Stem, 1.3mL Diameter (Metric) Outer 2.5 mm Material LDPE Sterility Sterile Drops/mL 26 Graduated No Packaging Type 20/Bag, 500/Pk., 4000/Cs. Volume (Metric) 1.3 mL Length (Metric) 6.4 cm Type Narrow Stem Transfer Pipet Disposable Single-use Unit Size Case of 4000
242 Full specifications
Case of 5000 4.3 mL 3.1 mL Non-sterile Bulk 500/Pk 10Pk/Cs 5000/Cs 418.71 Online price 670.00
- Bulb Draw (Metric) 3.1 mL Description Transfer Pipette, SEDI-PET, Short Stem, Long Bulb, 4.3mL Diameter (Metric) Outer 2.5 mm Material LDPE Sterility Non-sterile Drops/mL 24 Graduated No Packaging Type Bulk 500/Pk 10Pk/Cs 5000/Cs Volume (Metric) 4.3 mL Length (Metric) 8.5 cm Type Narrow Stem Transfer Pipet Disposable Single-use Unit Size Case of 5000
242PK also known as 242
Full specifications
Pack of 500 4.3 mL 3.1 mL Non-sterile Bulk 500/Pk 10Pk/Cs 5000/Cs - Bulb Draw (Metric) 3.1 mL Description Transfer Pipette, SEDI-PET, Short Stem, Long Bulb, 4.3mL Diameter (Metric) Outer 2.5 mm Material LDPE Sterility Non-sterile Drops/mL 24 Graduated No Packaging Type Bulk 500/Pk 10Pk/Cs 5000/Cs Volume (Metric) 4.3 mL Length (Metric) 8.5 cm Type Narrow Stem Transfer Pipet Disposable Single-use Unit Size Pack of 500
242-1S Full specifications
Case of 4000 4.3 mL 3.1 mL Sterile Individually Wrapped 500/Pk 8Pk/Cs 4000/Cs 948.65 Online price 1,191.00
- Bulb Draw (Metric) 3.1 mL Description Transfer Pipette, SEDI-PET, Short Stem, Long Bulb, 4.3mL Diameter (Metric) Outer 2.5 mm Material LDPE Sterility Sterile Drops/mL 24 Graduated No Packaging Type Individually Wrapped 500/Pk 8Pk/Cs 4000/Cs Volume (Metric) 4.3 mL Length (Metric) 8.5 cm Type Narrow Stem Transfer Pipet Disposable Single-use Unit Size Case of 4000
242-20S Full specifications
Case of 6080 4.3 mL 3.1 mL Sterile 20/Bag, 760/Pk., 6080/Cs. Contact Us
- - Bulb Draw (Metric) 3.1 mL Description Transfer Pipette, SEDI-PET, Short Stem, Long Bulb, 4.3mL Diameter (Metric) Outer 2.5 mm Material LDPE Sterility Sterile Drops/mL 24 Graduated No Packaging Type 20/Bag, 760/Pk., 6080/Cs. Volume (Metric) 4.3 mL Length (Metric) 8.5 cm Type Narrow Stem Transfer Pipet Disposable Single-use Unit Size Case of 6080
251 Full specifications
Case of 5000 4.5 mL 3.3 mL Non-sterile Bulk 500/Pk 10Pk/Cs 5000/Cs 296.56 Online price 486.00
- Bulb Draw (Metric) 3.3 mL Description Sedi-Pet Transfer Pipette, Narrow Stem, Standard Bulb 15.3mL Diameter (Metric) Outer 2.5 mm Material LDPE Sterility Non-sterile Drops/mL 26 Graduated No Packaging Type Bulk 500/Pk 10Pk/Cs 5000/Cs Volume (Metric) 4.5 mL Length (Metric) 15.5 cm Type Narrow Stem Transfer Pipet Disposable Single-use Unit Size Case of 5000
251PK also known as 251
Full specifications
Pack of 500 4.5 mL 3.3 mL Non-sterile Bulk 500/Pk 10Pk/Cs 5000/Cs - Bulb Draw (Metric) 3.3 mL Description Sedi-Pet Transfer Pipette, Narrow Stem, Standard Bulb 15.3mL Diameter (Metric) Outer 2.5 mm Material LDPE Sterility Non-sterile Drops/mL 26 Graduated No Packaging Type Bulk 500/Pk 10Pk/Cs 5000/Cs Volume (Metric) 4.5 mL Length (Metric) 15.5 cm Type Narrow Stem Transfer Pipet Disposable Single-use Unit Size Pack of 500