Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Thermo Scientific™ Nunc 384-Well Clear Polystyrene Plates with Cell Culture Treated and Nontreated Surfaces are optimized for robotics, plate readers and liquid handling systems. 384-well microplates allow sample and reagent volume reduction providing cost savings and allowing more samples to be screened. Our plates are high quality for consistent and reliable results.
Barcode | Custom Barcoding Available |
Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
Format | Standard |
Lid | With Lid |
Culture Area | 0.084 cm2/well |
Description | Non-Treated; w/ lid; Sterile; 10/30 |
Array | 16 x 24 |
Packaging | 10 Pk., 30 Cases |
Product Line | Nunc |
Color | Clear |
Product Type | Microplate |
Material | Polystyrene |
Volume (Metric) Well | 120 μL |
No. of Wells | 384 |
Volume (Metric) Working | 10 to 100 μL |
Sterility | Sterile |
Surface Treatment | Non-treated |
Well Shape | Flat Bottom (F-well) |
No. per Case | 30 |
No. per Pack | 10 |
Unit Size | Case of 30 |
Barcode | Custom Barcoding Available |
Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
Lid | With Lid |
Culture Area | 0.084 cm2/well |
Description | Non-Treated; w/ lid; Sterile; 1/30 |
Array | 16 x 24 |
Packaging | 1/Pk., 30/Cs. |
Product Line | Nunc |
Color | Clear |
Product Type | Microplate |
Material | Polystyrene |
Volume (Metric) Well | 120 μL |
No. of Wells | 96 |
Volume (Metric) Working | 10 to 100 μL |
Sterility | Sterile |
Surface Treatment | Non-treated |
Well Shape | Flat Bottom (F-well) |
No. per Case | 30 |
No. per Pack | 1 |
Unit Size | Case of 30 |
Barcode | Custom Barcoding Available |
Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
Lid | With Lid |
Culture Area | 0.084 cm2/well |
Description | Non-Treated; w/ lid; Sterile; 25/100 |
Array | 16 x 24 |
Packaging | 25/Pk., 100/Cs. |
Product Line | Nunc |
Color | Clear |
Product Type | Microplate |
Material | Polystyrene |
Volume (Metric) Well | 120 μL |
No. of Wells | 384 |
Volume (Metric) Working | 10 to 100 μL |
Sterility | Sterile |
Surface Treatment | Non-treated |
Well Shape | Flat Bottom (F-well) |
No. per Case | 100 |
No. per Pack | 25 |
Unit Size | Case of 100 |
Catalog Number | Specifications | Unit Size | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
242757 | Case of 30 | 244.65 Online price 257.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
242765 | Case of 30 | 312.65 Online price 328.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
265202 | Case of 100 | 727.65 Online price 764.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|