Thermo Scientific Chemicals

D-Lysine hydrochloride, 99+%

Catalog number: 243080500
50 g, Glass Bottle, Each
Thermo Scientific Chemicals

D-Lysine hydrochloride, 99+%

Catalog number: 243080500
50 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 243080500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7274-88-6
IUPAC Name
(2R)-2,6-diaminohexanoic acid hydrochloride
Molecular Formula
C6H15ClN2O2
InChI Key
BVHLGVCQOALMSV-XZNNNFJINA-N
SMILES
Cl.NCCCC[C@@H](N)C(O)=O
Appearance (Form)
Crystalline powder
Titration with HClO4
99.0 to 101.0 %
Specific optical rotation
-19° to -22° (20°C, 589 nm) (c=5, 5 N HCl)
Iron (Fe)
=<30 ppm
Appearance (Color)
White to almost white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text