Methyl 3-oxo-pentanoate, 99+%, Thermo Scientific Chemicals
Methyl 3-oxo-pentanoate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 3-oxo-pentanoate, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 243160250
Price (USD)/ Each
40.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
48.10 
Save 7.45 (15%)
-
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
40.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
48.10 
Save 7.45 (15%)
Add to cart
Methyl 3-oxo-pentanoate, 99+%, Thermo Scientific Chemicals
Catalog number243160250
Price (USD)/ Each
40.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
48.10 
Save 7.45 (15%)
-
Add to cart
Chemical Identifiers
CAS30414-53-0
IUPAC Namemethyl 3-oxopentanoate
Molecular FormulaC6H10O3
InChI KeyXJMIXEAZMCTAGH-UHFFFAOYSA-N
SMILESCCC(=O)CC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=99.0 %
Infrared spectrumConforms
Refractive index1.4220 to 1.4240 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only