Thermo Scientific Chemicals

Methyl 3-oxo-pentanoate, 99+%, Thermo Scientific Chemicals

Catalog number: 243161000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

Methyl 3-oxo-pentanoate, 99+%, Thermo Scientific Chemicals

Catalog number: 243161000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 243161000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
30414-53-0
IUPAC Name
methyl 3-oxopentanoate
Molecular Formula
C6H10O3
InChI Key
XJMIXEAZMCTAGH-UHFFFAOYSA-N
SMILES
CCC(=O)CC(=O)OC
Appearance (Form)
Liquid
GC
>=99.0 %
Infrared spectrum
Conforms
Refractive index
1.4220 to 1.4240 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text