Thermo Scientific Chemicals

Sodium, 30wt% dispersion in toluene, <0.1 mm particle size, Thermo Scientific Chemicals

Catalog number: 243262500
250 mL, Each
Thermo Scientific Chemicals

Sodium, 30wt% dispersion in toluene, <0.1 mm particle size, Thermo Scientific Chemicals

Catalog number: 243262500
250 mL, Each
Quantity
Catalog number: 243262500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7440-23-5
IUPAC Name
sodium
Molecular Formula
Na
InChI Key
KEAYESYHFKHZAL-UHFFFAOYSA-N
SMILES
[Na]
Appearance (Form)
Dispersion in toluene
Strength
>=30 % (Na)
Chloride (Cl)
=<50 ppm
Calcium (Ca)
=<550 ppm
Additional info
Trace analysis based on 100% sodium metal purity

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text