D-Calcium pantothenate, 98%
D-Calcium pantothenate, 98%
D-Calcium pantothenate, 98%
D-Calcium pantothenate, 98%
Thermo Scientific Chemicals

D-Calcium pantothenate, 98%

CAS: 137-08-6 | C18H32CaN2O10 | 476.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Glass Bottle
Catalog number 243300050
Price (USD)
37.65
Online Exclusive
41.40
Save 3.75 (9%)
Each
-
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
37.65
Online Exclusive
41.40
Save 3.75 (9%)
Each
Add to cart
Chemical Identifiers
CAS137-08-6
IUPAC Namecalcium bis(3-[(2R)-2,4-dihydroxy-3,3-dimethylbutanamido]propanoate)
Molecular FormulaC18H32CaN2O10
InChI KeyFAPWYRCQGJNNSJ-DXHDTSSINA-L
SMILES[Ca++].CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O.CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation+25.0° to +27.5° (20°C, 589 nm) (c=5, H2O)
Specific optical rotationOn dried substance
Loss on drying=<5.0 % (105°C, 3 h)
Heavy metals (as Pb)=<20 ppm
Appearance (Color)White to almost white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection