Thermo Scientific Chemicals

2-Hydroxypyridine-N-oxide, 97%, Thermo Scientific Chemicals

Catalog number: 243790250
25 g, Each
Thermo Scientific Chemicals

2-Hydroxypyridine-N-oxide, 97%, Thermo Scientific Chemicals

Catalog number: 243790250
25 g, Each
Quantity
Catalog number: 243790250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13161-30-3
IUPAC Name
1-hydroxy-1,2-dihydropyridin-2-one
Molecular Formula
C5H5NO2
InChI Key
SNUSZUYTMHKCPM-UHFFFAOYSA-N
SMILES
ON1C=CC=CC1=O
Appearance (Color)
Off-white to beige
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
147°C to 152°C
Titration with NaOH
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text