Kjeldahl tablets with selenium (Wieninger), Thermo Scientific Chemicals
Kjeldahl tablets with selenium (Wieninger), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Kjeldahl tablets with selenium (Wieninger), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
1 kg
Packaging:
Plastic bottle
Catalog number 243920025
Price (USD)/ Each
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Nameselenium
Molecular FormulaSe
InChI KeyBUGBHKTXTAQXES-UHFFFAOYSA-N
SMILES[Se]
Molecular Weight (g/mol)78.97
View more
SpecificationsSpecification SheetSpecification Sheet
Additional infoAverage weight range (per tablet): 4.85 to 5.15 g
Appearance (Color)Dark grey
Appearance (Form)Tablets with green or blue speckles
Nitrogen (N)=<0.05 %
Additional infoCopper sulphate pentahydrate (per tablet):
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only